Thermo Scientific Chemicals

Gold gauze, 82 mesh woven from 0.06mm (0.0025in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 040586.FF
25 x 25 mm, Each
Thermo Scientific Chemicals

Gold gauze, 82 mesh woven from 0.06mm (0.0025in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 040586.FF
25 x 25 mm, Each
Quantity
Catalog number: 040586.FF
also known as 040586-FF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-57-5
IUPAC Name
gold
Molecular Formula
Au
InChI Key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES
[Au]
Appearance (Color)
Golden
Form
Gauze, 82 mesh woven from 0.06mm (0.0025in) diameter wire.
Assay from Supplier's CofA
≥99.9% (metals basis)

Description

Gold gauze is used to trap mercury vapors during the analysis of mercury by cold vapor-atomic absorption spectrophotometer (CVAAS). It is also used as a surface for condensation of mercury. Further, it is used as an electrode in electrochemistry for the reduction of uranium(V).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text