Boron nitride, Thermo Scientific Chemicals
Boron nitride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron nitride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 040607.04
also known as 040607-04
Price (USD)/ Each
106.00
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
106.00
Add to cart
Boron nitride, Thermo Scientific Chemicals
Catalog number040607.04
Price (USD)/ Each
106.00
-
Add to cart
Chemical Identifiers
CAS10043-11-5
IUPAC Nameboranimine
Molecular FormulaBN
InChI KeyLNLSXDSWJBUPHM-UHFFFAOYSA-N
SMILESB=N
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size4-8 Microns
Boron nitride is used as polymorph, cosmetics, paints, dental cements, and pencil leads. It is also used in xerographic process and laser printers as a charge leakage barrier layer of the photo drum, fillers, processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only