Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 040619.14
also known as 040619-14
Price (USD)/ Each
29.65
Online exclusive
32.30 
Save 2.65 (8%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
29.65
Online exclusive
32.30 
Save 2.65 (8%)
Add to cart
Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Catalog number040619.14
Price (USD)/ Each
29.65
Online exclusive
32.30 
Save 2.65 (8%)
-
Add to cart
Chemical Identifiers
CAS28300-74-5
IUPAC Namedipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
Molecular FormulaC8H10K2O15Sb2
InChI KeyWBTCZEPSIIFINA-UHFFFAOYNA-J
SMILESO.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder or powder
Assay (Iodometric Titration)99.0 to 103.0% [K2(C4H2O6Sb)2.3H2O]
Comment≤0.020 meq/g (Titrable free acid or base)
Impurity content≤0.015% Arsenic (As)
View more
Antimony potassium tartrate trihydrate is used as a antiparasitic agent, which used in the treatment of schistosomiasis and leishmaniasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only