Thermo Scientific Chemicals

Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Catalog number: 040619.36
500 g, Each
Thermo Scientific Chemicals

Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Catalog number: 040619.36
500 g, Each
Quantity
Catalog number: 040619.36
also known as 040619-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28300-74-5
IUPAC Name
dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
Molecular Formula
C8H10K2O15Sb2
InChI Key
WBTCZEPSIIFINA-UHFFFAOYNA-J
SMILES
O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
Appearance (Color)
White
Form
Crystalline powder or powder
Assay (Iodometric Titration)
99.0 to 103.0% [K2(C4H2O6Sb)2.3H2O]
Comment
≤0.020 meq/g (Titrable free acid or base)
Impurity content
≤0.015% Arsenic (As)

Description

Antimony potassium tartrate trihydrate is used as a antiparasitic agent, which used in the treatment of schistosomiasis and leishmaniasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text