Hafnium(IV) oxide, 99.99% (metals basis excluding Zr), Zr <100ppm, Thermo Scientific Chemicals
Hafnium(IV) oxide, 99.99% (metals basis excluding Zr), Zr &lt;100ppm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium(IV) oxide, 99.99% (metals basis excluding Zr), Zr <100ppm, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040620.06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS12055-23-1
IUPAC Namehafnium(4+) dioxidandiide
Molecular FormulaHfO2
InChI KeyWIHZLLGSGQNAGK-UHFFFAOYSA-N
SMILES[O--].[O--].[Hf+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentZr <100ppm
Total Metal Impurities excluding Zr0.01% max.
Hafnium(IV) oxide is used as Intermediates, Paint additives and coating additives, metal Products. And it is also used in optical coatings, as a refractory material in the insulation of such devices as thermocouples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only