Yttrium(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Yttrium(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 040651.30
also known as 040651-30
Price (USD)/ Each
305.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
305.00
Add to cart
Yttrium(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number040651.30
Price (USD)/ Each
305.00
-
Add to cart
Chemical Identifiers
CAS38245-39-5
IUPAC Namediyttrium(3+) tricarbonate
Molecular FormulaC3O9Y2
InChI KeyQVOIJBIQBYRBCF-UHFFFAOYSA-H
SMILES[Y+3].[Y+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only