Thermo Scientific Chemicals

Ytterbium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 040653.14
25 g, Each
Thermo Scientific Chemicals

Ytterbium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 040653.14
25 g, Each
Quantity
Catalog number: 040653.14
also known as 040653-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10361-91-8
IUPAC Name
ytterbium(3+) trichloride
Molecular Formula
Cl3Yb
InChI Key
CKLHRQNQYIJFFX-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Yb+3]
Appearance (Color)
White
Form
Beads
Residual water
< 100 ppm
Total Rare Earth Oxide Impurities
0.01% max.
Particle Size
-10 Mesh

Description

Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides. Ytterbium(III) chloride is a powerful catalyst for the formation of acetals using trimethyl orthoformate and also used to track digestion in animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text