Thermo Scientific Chemicals

Lithium iodide, anhydrous, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 040666.14
25 g, Each
Thermo Scientific Chemicals

Lithium iodide, anhydrous, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 040666.14
25 g, Each
Quantity
Catalog number: 040666.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10377-51-2
IUPAC Name
lithium(1+) iodide
Molecular Formula
ILi
InChI Key
HSZCZNFXUDYRKD-UHFFFAOYSA-M
SMILES
[Li+].[I-]
Appearance (Color)
Pale gray
Form
Crystalline/Powder
Total Metal Impurities
≤0.05% (500 ppm)

Description

Used as an electrolyte in high temperature batteries and artificial pacemakers. Phosphor , which is a complex of Iodine is used in neuron detection , in an electrolyte of the dye-sensitized solar cell. It is an electrolyte additive for lithium-sulfur batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text