Strontium 2-ethylhexanoate, 40% in 2-ethylhexanoic acid (8-12% Sr), Thermo Scientific Chemicals
Strontium 2-ethylhexanoate, 40% in 2-ethylhexanoic acid (8-12% Sr), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium 2-ethylhexanoate, 40% in 2-ethylhexanoic acid (8-12% Sr), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 040719.14
also known as 040719-14
Price (USD)/ Each
171.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
171.00
Add to cart
Strontium 2-ethylhexanoate, 40% in 2-ethylhexanoic acid (8-12% Sr), Thermo Scientific Chemicals
Catalog number040719.14
Price (USD)/ Each
171.00
-
Add to cart
Specifications
Chemical Name or MaterialStrontium 2-ethylhexanoate
CAS2457-02-5
Health Hazard 1H361d
Health Hazard 2GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Health Hazard 3P201-P202-P281-P308+P313-P501c
View more
Strontium 2-ethylhexanoate is used in metal refining, plumbing, bleaching, engraving, plating, photography, disinfection, fertilizer manufacture, metal cleaning, and rust removal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only