Copper(I) bromide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Copper(I) bromide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(I) bromide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 040752.03
also known as 040752-03
Price (USD)/ Each
83.65
Online exclusive
92.50 
Save 8.85 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
83.65
Online exclusive
92.50 
Save 8.85 (10%)
Add to cart
Copper(I) bromide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number040752.03
Price (USD)/ Each
83.65
Online exclusive
92.50 
Save 8.85 (10%)
-
Add to cart
Chemical Identifiers
CAS7787-70-4
IUPAC Nameλ¹-copper(1+) bromide
Molecular FormulaBrCu
InChI KeyNKNDPYCGAZPOFS-UHFFFAOYSA-M
SMILES[Cu+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
It is used as effective catalyst for tetrahydropyranylation of alcohols. It can also be used in combination with palladium in catalytic synthesis of 3-haloindoles through annulation process. It is widely used in the synthesis of organic fine chemicals. The compound is widely used in the synthesis of organic compounds. It is also useful for copper-mediated coupling reactions. A silica gel support copper bromide hexa methyltriethylenetetramine complex has shown useful application for the atom transfer radical polymerization of methyl methacrylate in toluene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only