Thermo Scientific Chemicals

Copper(I) bromide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 040752.06
5 g, Each
Thermo Scientific Chemicals

Copper(I) bromide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 040752.06
5 g, Each
Quantity
Catalog number: 040752.06
also known as 040752-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7787-70-4
IUPAC Name
λ¹-copper(1+) bromide
Molecular Formula
BrCu
InChI Key
NKNDPYCGAZPOFS-UHFFFAOYSA-M
SMILES
[Cu+].[Br-]
Total Metal Impurities
0.002% max.

Description

It is used as effective catalyst for tetrahydropyranylation of alcohols. It can also be used in combination with palladium in catalytic synthesis of 3-haloindoles through annulation process. It is widely used in the synthesis of organic fine chemicals. The compound is widely used in the synthesis of organic compounds. It is also useful for copper-mediated coupling reactions. A silica gel support copper bromide hexa methyltriethylenetetramine complex has shown useful application for the atom transfer radical polymerization of methyl methacrylate in toluene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text