Thermo Scientific Chemicals

Lead(II) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 040753.36
500 g, Each
Thermo Scientific Chemicals

Lead(II) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 040753.36
500 g, Each
Quantity
Catalog number: 040753.36
also known as 040753-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1317-36-8
IUPAC Name
plumbanone
Molecular Formula
OPb
InChI Key
YEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILES
O=[Pb]
Total Metal Impurities
0.001% max.

Description

Used in producing iridescent colors on brass and bronze, coloring sulfur-containing substances and used as pigment for rubber. It is also used in oil refining, varnishes, paints, enamels and in assays for precious metal ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text