Magnesium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals
Magnesium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 040755.04
also known as 040755-04
Price (USD)/ Each
70.50
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
70.50
Add to cart
Magnesium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number040755.04
Price (USD)/ Each
70.50
-
Add to cart
Chemical Identifiers
CAS7783-40-6
IUPAC Namemagnesium(2+) difluoride
Molecular FormulaF2Mg
InChI KeyORUIBWPALBXDOA-UHFFFAOYSA-L
SMILES[F-].[F-].[Mg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Used in optics, space telescopes. Most commonly used on the surfaces of optical elements as inexpensive anti-reflective coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only