Thermo Scientific Chemicals

Manganese(II) carbonate, Puratronic™, 99.985% (metals basis), Thermo Scientific Chemicals

Catalog number: 040756.14
25 g, Each
Thermo Scientific Chemicals

Manganese(II) carbonate, Puratronic™, 99.985% (metals basis), Thermo Scientific Chemicals

Catalog number: 040756.14
25 g, Each
Quantity
Catalog number: 040756.14
also known as 040756-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
598-62-9
IUPAC Name
manganese(2+) carbonate
Molecular Formula
CMnO3
InChI Key
XMWCXZJXESXBBY-UHFFFAOYSA-L
SMILES
[Mn++].[O-]C([O-])=O
Total Metal Impurities
0.0015% max.

Description

Manganese(II) carbonate is used in the production of manganese dioxide for dry-cell batteries and for ferrites, as an additive to plant fertilizers to cure manganese deficient crops. It is also used in health foods, in ceramics as a glaze colorant and flux, and in concrete stains, and as a hematinic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text