Manganese(II) carbonate, Puratronic™, 99.985% (metals basis), Thermo Scientific Chemicals
Manganese(II) carbonate, Puratronic™, 99.985% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) carbonate, Puratronic™, 99.985% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 040756.22
also known as 040756-22
Price (USD)/ Each
577.65
Online exclusive
642.00 
Save 64.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
577.65
Online exclusive
642.00 
Save 64.35 (10%)
Add to cart
Manganese(II) carbonate, Puratronic™, 99.985% (metals basis), Thermo Scientific Chemicals
Catalog number040756.22
Price (USD)/ Each
577.65
Online exclusive
642.00 
Save 64.35 (10%)
-
Add to cart
Chemical Identifiers
CAS598-62-9
IUPAC Namemanganese(2+) carbonate
Molecular FormulaCMnO3
InChI KeyXMWCXZJXESXBBY-UHFFFAOYSA-L
SMILES[Mn++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0015% max.
Manganese(II) carbonate is used in the production of manganese dioxide for dry-cell batteries and for ferrites, as an additive to plant fertilizers to cure manganese deficient crops. It is also used in health foods, in ceramics as a glaze colorant and flux, and in concrete stains, and as a hematinic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only