Aluminum foil, 2.0mm (0.08in) thick, annealed, 99.99% (metals basis), Thermo Scientific Chemicals
Aluminum foil, 2.0mm (0.08in) thick, annealed, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum foil, 2.0mm (0.08in) thick, annealed, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
50 x 50 mm
100 x 500 mm
Catalog number 040763.GH
also known as 040763-GH
Price (USD)/ Each
212.00
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
212.00
Add to cart
Aluminum foil, 2.0mm (0.08in) thick, annealed, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number040763.GH
Price (USD)/ Each
212.00
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 2.0mm (0.08in) thick, annealed.
Assay from Supplier's CofA≥99.99% (metals basis)
Appearance (Color)Silver
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only