Graphite rod, 3.05mm (0.12in) dia x 305mm (12in) long, 99.9995% (metals basis), Thermo Scientific Chemicals
Graphite rod, 3.05mm (0.12in) dia x 305mm (12in) long, 99.9995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Graphite rod, 3.05mm (0.12in) dia x 305mm (12in) long, 99.9995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 Pc.
25 Pc.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040765.KN
Price (USD)/ Each
-
Quantity:
100 Pc.
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialGraphite rod
Name Note3.05mm (0.12 in.) dia. x 305mm (12 in.) long
Recommended StorageAmbient temperatures
EINECS Number231-955-3
TSCAYes
View more
Graphite rod is used in computer brushes, anodes, arc lamps, carbon coating, cathode ray tubes, and moderator in nuclear piles. It is used for high-temperature crucibles, as a lubricant and in lead pencils. Graphite rod coated with Th(IV), plays an useful application for ion selective electrode based on thorium oxinate complex. It is also essential for the preparation of lead(II)-selective PVC-coated graphite rod electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Commutar brushes, anodes, arc lamps, carbon coating, cathode ray tubes, moderator in nuclear piles
RUO – Research Use Only