Thermo Scientific Chemicals

Graphite powder, natural, briquetting grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 040795.TC
227 g, Each
Thermo Scientific Chemicals

Graphite powder, natural, briquetting grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 040795.TC
227 g, Each
Quantity
Catalog number: 040795.TC
Price (USD)
Have Questions?
Assay from Supplier's CofA
≥99.9995% (metals basis)
Particle Size
ca. 200 Mesh
Appearance (Color)
Grey

Description

Graphite Powder is basically used for casting and polishing purposes, moulding & Carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text