Thermo Scientific Chemicals

Polypropylene glycol 400, Thermo Scientific Chemicals

Catalog number: 040811.18
50 g, Each
Thermo Scientific Chemicals

Polypropylene glycol 400, Thermo Scientific Chemicals

Catalog number: 040811.18
50 g, Each
Quantity
Catalog number: 040811.18
also known as 040811-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25322-69-4
Molecular Formula
(C3H6O)n
InChI Key
DNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES
[H]OC(C)CO
MDL Number
MFCD00084446
Form
viscous liquid
Appearance (Color)
Colorless

Description

It is used in cosmetic and pharmaceutical preparations and in the manufacture of emulsifying or wetting agents and lubricants,suppress foaming in industrial processes and for making polyurethane resins, hydraulic fluids, and various other materials. It acts as swelling agent for the preparation and characterization of organo-modified SBA-15. Polypropylene glycol and tetraethoxysilane precursors play an important role for the preparation of superhydrophobic silica-based surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text