Polypropylene glycol 3,000, Thermo Scientific Chemicals
Polypropylene glycol 3,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polypropylene glycol 3,000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 040814.30
also known as 040814-30
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
Add to cart
Polypropylene glycol 3,000, Thermo Scientific Chemicals
Catalog number040814.30
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS25322-69-4
Molecular Formula(C3H6O)n
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES[H]OC(C)CO
MDL NumberMFCD00084446
SpecificationsSpecification SheetSpecification Sheet
CommentMolecular Weight: Approximately 3000
Hydroxyl ValueReport Value
Viscosity400-700 cs
FormLiquid
Propylene glycol is used in the plastics industry for the manufacture of polyester fibers and alkyd resins. It is used as a main ingredient in automobile antifreeze and engine-cooling liquids and in brake and hydraulic fluids. It is used as a humectant in foods and cosmetics. It is used as a solvent for coloring or flavoring agents as well as in many oral, injectable or topical pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only