Thermo Scientific Chemicals

Germanium n-butoxide, 97+%, Thermo Scientific Chemicals

Catalog number: 040816.03
1 g, Each
Thermo Scientific Chemicals

Germanium n-butoxide, 97+%, Thermo Scientific Chemicals

Catalog number: 040816.03
1 g, Each
Quantity
Catalog number: 040816.03
also known as 040816-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25063-27-8
Molecular Formula
C16H32GeO4
InChI Key
MPVCRVXMJOZDGL-UHFFFAOYSA-N
SMILES
[Ge].[CH2]CCC[O-].[CH2]CCC[O-].[CH2]CCC[O-].[CH2]CCC[O-]
Molecular Weight (g/mol)
361.06
Infrared spectrum
Conforms
Assay from Supplier's CofA
≥97%
Appearance (Color)
Clear colorless
Form
Liquid

Description

Germanium n-butoxide is used as oxide films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text