Copper sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.999% (metals basis), Thermo Scientific Chemicals
Copper sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.999% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Ea.
Catalog number 040839.KS
Price (USD)/ Each
465.00
-
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
465.00
Add to cart
Copper sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number040839.KS
Price (USD)/ Each
465.00
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormSputtering target, 50.8mm (2.0in) diameter x 6.35mm (0.250in) thick.
Assay from Supplier's CofA≥99.999% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only