Iron sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Iron sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Ea.
Catalog number 040855.KS
Price (USD)/ Each
417.00
-
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
417.00
Add to cart
Iron sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number040855.KS
Price (USD)/ Each
417.00
-
Add to cart
Specifications
Chemical Name or MaterialIron sputtering target
Name Note50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Quantity1 Ea.
CAS7439-89-6
Health Hazard 1H500
View more
Iron sputtering target is used in semiconductor, chemical vapor deposition (CVD) and physical vapor deposition (PVD) display and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only