Manganese sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Manganese sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Ea.
Catalog number 040863.KS
Price (USD)/ Each
1,176.00
-
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
1,176.00
Add to cart
Manganese sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number040863.KS
Price (USD)/ Each
1,176.00
-
Add to cart
Specifications
Chemical Name or MaterialManganese sputtering target
Name Note50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Quantity1 Ea.
CAS7439-96-5
FormHot Pressed
View more
Manganese sputtering target is use in optical display , semiconductor, chemical vapor deposition and physical vapor deposition (PVD) and other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only