Molybdenum sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Molybdenum sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Ea.
Catalog number 040869.KS
Price (USD)/ Each
208.00
-
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
208.00
Add to cart
Molybdenum sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number040869.KS
Price (USD)/ Each
208.00
-
Add to cart
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
Assay from Supplier's CofA≥99.95% (metals basis)
FormSputtering target, 76.2mm (3.0in) diameter x 6.35mm (0.250in) thick.
The demand for molybdenum sputtering target increases with the rapid development of the flat panel display industry and photovoltaic industry. The as-rolled and recrystallized molybdenum plates were made to sputter thin films and to examine the effects of sputtering target microstructures upon sputtered films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only