Nickel sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Nickel sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Ea.
Catalog number 040874.KS
Price (USD)/ Each
315.65
Online exclusive
350.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
315.65
Online exclusive
350.00 
Save 34.35 (10%)
Add to cart
Nickel sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number040874.KS
Price (USD)/ Each
315.65
Online exclusive
350.00 
Save 34.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery white
Assay from Supplier's CofA≥99.99% (metals basis)
FormSputtering target, 50.8mm (2.0in) diameter x 3.18mm (0.125in) thick.
Magnetron sputtering source

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only