Nickel sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Nickel sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Ea.
Catalog number 040876.KS
Price (USD)/ Each
465.00
-
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
465.00
Add to cart
Nickel sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number040876.KS
Price (USD)/ Each
465.00
-
Add to cart
Specifications
Chemical Name or MaterialNickel sputtering target
Name Note76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Quantity1 Ea.
CAS7440-02-0
Health Hazard 1H317-H351-H372
View more
The accumulation of radiation defects in the crystal lattice of nickel in the course of irradiation leads to an increase in the metal sputtering yield from the surface of grains, making it comparable with the sputtering yield from the intergranular region. These result in development of the heavy ion acceleration facilities and for the high-energy heavy-ion doping of deep layers in semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only