Niobium sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis excluding Ta), Thermo Scientific Chemicals
Niobium sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis excluding Ta), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis excluding Ta), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Ea.
Catalog number 040883.KS
Price (USD)/ Each
443.00
-
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
443.00
Add to cart
Niobium sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis excluding Ta), Thermo Scientific Chemicals
Catalog number040883.KS
Price (USD)/ Each
443.00
-
Add to cart
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.95% (metals basis excluding Ta)
Appearance (Color)Gray
FormSputtering target, 50.8mm (2.0in) diameter x 6.35mm (0.250in) thick.
Niobium sputtering target is used in high temperature engineering products. It is also used as an alloying agent for certain steels where it greatly improves the strength of the resulting material. It is also finds applications in atomic reactors due to its corrosion resistance and, when combined with either tin (Nb3Sn) or zirconium, it has a high degree of superconductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only