Molybdenum gauze, 100 mesh woven from 0.0636mm (0.0025in) dia wire, Thermo Scientific Chemicals
Molybdenum gauze, 100 mesh woven from 0.0636mm (0.0025in) dia wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum gauze, 100 mesh woven from 0.0636mm (0.0025in) dia wire, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
300 x 300 mm
75 x 75 mm
150 x 150 mm
Catalog number 040912.HB
also known as 040912-HB
Price (USD)/ Each
1,456.00
-
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (USD)/ Each
1,456.00
Add to cart
Molybdenum gauze, 100 mesh woven from 0.0636mm (0.0025in) dia wire, Thermo Scientific Chemicals
Catalog number040912.HB
Price (USD)/ Each
1,456.00
-
Add to cart
Specifications
Chemical Name or MaterialMolybdenum gauze
Name NoteWoven from 0.0636mm (0.0025 in.) dia. wire
CAS7439-98-7
Recommended StorageAmbient temperatures
EINECS Number231-107-2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only