Titanium gauze, 20 mesh woven from 0.076mm (0.003 in.) dia. wire, Thermo Scientific Chemicals
Titanium gauze, 20 mesh woven from 0.076mm (0.003 in.) dia. wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium gauze, 20 mesh woven from 0.076mm (0.003 in.) dia. wire, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
300 x 300 mm
75 x 75 mm
Catalog number 040918.HB
also known as 040918-HB
Price (USD)/ Each
688.00
-
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (USD)/ Each
688.00
Add to cart
Titanium gauze, 20 mesh woven from 0.076mm (0.003 in.) dia. wire, Thermo Scientific Chemicals
Catalog number040918.HB
Price (USD)/ Each
688.00
-
Add to cart
Specifications
Chemical Name or MaterialTitanium gauze
Name NoteWoven from 0.076mm (0.003 in.) dia. wire
CAS7440-32-6
Recommended StorageAmbient temperatures
EINECS Number231-142-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only