Titanium gauze, 40 mesh woven from 0.127mm (0.005in) dia wire, Thermo Scientific Chemicals
Titanium gauze, 40 mesh woven from 0.127mm (0.005in) dia wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium gauze, 40 mesh woven from 0.127mm (0.005in) dia wire, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
300 x 300 mm
75 x 75 mm
150 x 150 mm
Catalog number 040920.HB
also known as 040920-HB
Price (USD)/ Each
499.65
Online exclusive
555.00 
Save 55.35 (10%)
-
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (USD)/ Each
499.65
Online exclusive
555.00 
Save 55.35 (10%)
Add to cart
Titanium gauze, 40 mesh woven from 0.127mm (0.005in) dia wire, Thermo Scientific Chemicals
Catalog number040920.HB
Price (USD)/ Each
499.65
Online exclusive
555.00 
Save 55.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Metallic grey
FormGauze, 40 mesh woven from 0.005 in. diameter wire
CommentTitanium: ≥99.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only