Thermo Scientific Chemicals
Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number: 040930.FY
50 x 100 mm, Each
Thermo Scientific Chemicals
Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number: 040930.FY
50 x 100 mm, Each
Quantity
Catalog number: 040930.FY
also known as 040930-FY
Price (USD)
Quantity
-
Have Questions?
Description
Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis) is used as an electrode.