Silver gauze, 80 mesh woven from 0.115mm (0.0045in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silver gauze, 80 mesh woven from 0.115mm (0.0045in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
300 x 300 mm
25 x 25 mm
75 x 75 mm
150 x 150 mm
Catalog number 040936.HB
also known as 040936-HB
Price (USD)/ Each
1,898.00
-
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (USD)/ Each
1,898.00
Add to cart
Silver gauze, 80 mesh woven from 0.115mm (0.0045in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number040936.HB
Price (USD)/ Each
1,898.00
-
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment80 mesh woven from 0.115mm dia wire
Appearance (Color)Silver
Total Metal Impurities≤ 0.1%
FormGauze
Silver gauze is used for filtration. It is also used in electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only