Stainless Steel wire, 0.127mm (0.005in) dia, Type 304, Thermo Scientific Chemicals
Stainless Steel wire, 0.127mm (0.005in) dia, Type 304, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stainless Steel wire, 0.127mm (0.005in) dia, Type 304, Thermo Scientific Chemicals

Filtration, electrodes
Have Questions?
Change viewbuttonViewtableView
Quantity:
150 cm
50 cm
Catalog number 040944.BZ
also known as 040944-BZ
Price (USD)/ Each
47.65
Online exclusive
53.10 
Save 5.45 (10%)
-
Add to cart
Quantity:
150 cm
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
53.10 
Save 5.45 (10%)
Add to cart
Stainless Steel wire, 0.127mm (0.005in) dia, Type 304, Thermo Scientific Chemicals
Catalog number040944.BZ
Price (USD)/ Each
47.65
Online exclusive
53.10 
Save 5.45 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialStainless Steel wire, Type 304
Name Note0.127mm (0.005 in.) dia.
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only