Thermo Scientific Chemicals

Chloroform, Environmental Grade, 99.8+%, stab. with ethanol, Thermo Scientific Chemicals

Catalog number: 040974.M1
4 x 1 L, Each
Thermo Scientific Chemicals

Chloroform, Environmental Grade, 99.8+%, stab. with ethanol, Thermo Scientific Chemicals

Catalog number: 040974.M1
4 x 1 L, Each
Quantity
Catalog number: 040974.M1
also known as 040974-M1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
67-66-3
IUPAC Name
trichloromethane
Molecular Formula
CHCl3
InChI Key
HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES
ClC(Cl)Cl
Residual halide
10 ng/l (GC-ECD)
Residue on Evaporation
2ppm

Description

It is suitable for pesticide analysis, environmental and GC analysis. It is a commonly used laboratory solvent. It was previously used as an anesthetic. It is also used as a precursor to PTFE and refrigerants. Chloroform was used in the past as an extraction solvent for fats, oils and greases. It is useful for the rapid and simple release of periplasmic proteins. The mixture of guanidinium thiocyanate-phenol-chloroform, plays an essential role in the RNA isolation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text