Thermo Scientific Chemicals

Ethyl acetate, Environmental Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 040977.K2
1 L, Each
Thermo Scientific Chemicals

Ethyl acetate, Environmental Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 040977.K2
1 L, Each
Quantity
Catalog number: 040977.K2
also known as 040977-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-78-6
IUPAC Name
ethyl acetate
Molecular Formula
C4H8O2
InChI Key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES
CCOC(C)=O
Assay (unspecified)
10 ppt as Heptachloroepoxide (GC-ECD responsive residue as a single peak)
Residue on Evaporation
5ppm, max
Assay (GC)
99.5%, min
Residual water
0.05% max

Description

Ethyl acetate is used primarily as a solvent and diluent. It is commonly used to clean the circuit boards, in some nail varnish removers and also in residue, environmental and GC analysis. It is used chiefly as a scent in the manufacture of perfumes. It is also used in flavorings, glues, in decaffeinating tea and coffee, and in cigarettes. The other minor applications of ethyl acetate include its applications in the textiles, dyestuffs, explosives, photographic films and plates, artificial leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text