n-Heptane, Environmental Grade, 96+%, Thermo Scientific Chemicals
n-Heptane, Environmental Grade, 96+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Heptane, Environmental Grade, 96+%, Thermo Scientific Chemicals

For Pesticide Residue, Environmental, and GC Analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number 040978.K7
also known as 040978-K7
Price (USD)/ Each
147.65
Online exclusive
164.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
147.65
Online exclusive
164.00 
Save 16.35 (10%)
Add to cart
n-Heptane, Environmental Grade, 96+%, Thermo Scientific Chemicals
Catalog number040978.K7
Price (USD)/ Each
147.65
Online exclusive
164.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless, ≤10 APHA
Assay (GC)GC-FID ≥96.0% (n-Heptane)
Specific Absorbance (E 1%/1cm)197 nm ≤ 1.00 AU
Specific Absorbance (E 1%/1cm)225 nm ≤ 0.10 AU
Specific Absorbance (E 1%/1cm)280 nm ≤ 0.01 AU
View more
It is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing.used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Submicron filtered, halogenated residue: 10ng/L (GC-ECD)
RUO – Research Use Only