n-Pentane, Environmental Grade, 98+%, Thermo Scientific Chemicals
n-Pentane, Environmental Grade, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Pentane, Environmental Grade, 98+%, Thermo Scientific Chemicals

Suitable For Environmental, Pesticide, and GC Analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 4 L
Catalog number 040981.K2
also known as 040981-K2
Price (USD)/ Each
69.70
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
69.70
Add to cart
n-Pentane, Environmental Grade, 98+%, Thermo Scientific Chemicals
Catalog number040981.K2
Price (USD)/ Each
69.70
-
Add to cart
Chemical Identifiers
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)GC-FID ≥99.8% (C5 Hydrocarbons)
Water Content (Karl Fischer Titration)≤0.02%
Appearance (Color)Clear colorless, ≤10 APHA
Assay from Supplier's CofAGC-ECD Responsive residue (single peak) (as Heptachlorepoxide) ≤10 ppt
Residue on Evaporation≤5ppm
View more
n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only