Thermo Scientific Chemicals

2-Propanol, GC Grade, 99.7+%, Thermo Scientific Chemicals

Catalog number: 040983.K7
4 L, Each
Thermo Scientific Chemicals

2-Propanol, GC Grade, 99.7+%, Thermo Scientific Chemicals

Catalog number: 040983.K7
4 L, Each
Quantity
Catalog number: 040983.K7
also known as 040983-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-63-0
IUPAC Name
propan-2-ol
Molecular Formula
C3H8O
InChI Key
KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES
CC(C)O
Residue on Evaporation
≤5ppm
Form
Liquid
Color (APHA)
≤10
Assay from Suppliers CofA
≥99.7%
Residual water
≤0.10%

Description

Suitable for environmental and GC analysis. Used in making cosmetics, skin and hair preparations, pharmaceuticals, perfumes, lacquer formulations, dye solutions, anti freezes, soaps and window cleaners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text