Amberlite™ IRN-150, ion exchange resin, Thermo Scientific Chemicals
Amberlite™ IRN-150, ion exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amberlite™ IRN-150, ion exchange resin, Thermo Scientific Chemicals

Styrene-DVB; Mix of strongly acidic and basic gel type resin
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
500 g
2 kg
Catalog number 040992.A9
Price (USD)/ Each
444.65
Online exclusive
494.00 
Save 49.35 (10%)
-
Add to cart
Quantity:
10 kg
Request bulk or custom format
Price (USD)/ Each
444.65
Online exclusive
494.00 
Save 49.35 (10%)
Add to cart
Amberlite™ IRN-150, ion exchange resin, Thermo Scientific Chemicals
Catalog number040992.A9
Price (USD)/ Each
444.65
Online exclusive
494.00 
Save 49.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialAmberlite IRN-150
Physical FormBeads
Health Hazard 1H318
Health Hazard 3P280-P305+P351+P338-P310
Recommended StorageAmbient temperatures
View more
Amberlite IRN-150 is used in water treatment and purification in primary side chemical volume control systems (CVS) in PWR nuclear power generating facilities. It is also used in reactor water clean-up and radio active waste systems. It is recommended in any non-regenerable mixed bed application and can also be used for a variety of electroplating industry waste regeneration applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only