Thermo Scientific Chemicals

cis-Dichlorobis(triphenylphosphine)platinum(II), Premion™, 99.95% (metals basis), Pt 24.2% min, Thermo Scientific Chemicals

Catalog number: 041008.06
5 g, Each
Thermo Scientific Chemicals

cis-Dichlorobis(triphenylphosphine)platinum(II), Premion™, 99.95% (metals basis), Pt 24.2% min, Thermo Scientific Chemicals

Catalog number: 041008.06
5 g, Each
Quantity
Catalog number: 041008.06
also known as 041008-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15604-36-1
IUPAC Name
dichloroplatinumbis(ylium); bis(triphenylphosphane)
Molecular Formula
C36H30Cl2P2Pt
InChI Key
ILIBUWCDNDIUOT-UHFFFAOYSA-L
SMILES
Cl[Pt++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Total Metal Impurities
0.05% max., Pt 24.2% min

Description

cis-Dichlorobis(triphenylphosphine)platinum(II) is used in hydrosilylation, wax oil catalytic cracking, platinum plating and as platinum catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text