Thermo Scientific Chemicals

Potassium hexahydroxyplatinate(IV), Premion™, 99.95% (metals basis), Pt 51.5% min, Thermo Scientific Chemicals

Catalog number: 041014.06
5 g, Each
Thermo Scientific Chemicals

Potassium hexahydroxyplatinate(IV), Premion™, 99.95% (metals basis), Pt 51.5% min, Thermo Scientific Chemicals

Catalog number: 041014.06
5 g, Each
Quantity
Catalog number: 041014.06
also known as 041014-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12285-90-4
IUPAC Name
platinum(4+) dipotassium hexahydroxide
Molecular Formula
H6K2O6Pt
InChI Key
ALHHYLLHZKAXCW-UHFFFAOYSA-H
SMILES
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[K+].[K+].[Pt+4]
Total Metal Impurities
0.05% max.
Assay (unspecified)
Pt 51.5% min

Description

Potassium hexahydroxyplatinate(IV) is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text