Thermo Scientific Chemicals

Rhodium(III) sulfate tetrahydrate, Premion™, 99.99% (metals basis), Rh 35.9% min, Thermo Scientific Chemicals

Catalog number: 041016.03
1 g, Each
Thermo Scientific Chemicals

Rhodium(III) sulfate tetrahydrate, Premion™, 99.99% (metals basis), Rh 35.9% min, Thermo Scientific Chemicals

Catalog number: 041016.03
1 g, Each
Quantity
Catalog number: 041016.03
also known as 041016-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15274-78-9
IUPAC Name
dirhodium(3+) tetrahydrate trisulfate
Molecular Formula
H8O16Rh2S3
InChI Key
JJITWKYISWZJBI-UHFFFAOYSA-H
SMILES
O.O.O.O.[Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Total Metal Impurities
0.01% max., Rh 35.9% min

Description

Rhodium(III) sulfate is used as plating reagents and additives for noble metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text