Thermo Scientific Chemicals
Lead Tin, solder alloy, 3.18mm (0.125in) dia, Thermo Scientific Chemicals
Catalog number: 041023.29
225 g, Each
Thermo Scientific Chemicals
Lead Tin, solder alloy, 3.18mm (0.125in) dia, Thermo Scientific Chemicals
Catalog number: 041023.29
225 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 041023.29
also known as 041023-29
Price (USD)
Quantity
-
Have Questions?
Description
For use with copper, copper alloys, lead, steel, and nickel alloys. Recommended for light electrical wiring
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov