Zirconium(IV) sulfate tetrahydrate, 98+% (metals basis), Thermo Scientific Chemicals
Zirconium(IV) sulfate tetrahydrate, 98+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) sulfate tetrahydrate, 98+% (metals basis), Thermo Scientific Chemicals

Catalyst support
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041041.36
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7446-31-3
IUPAC Namezirconium(4+) tetrahydrate disulfate
Molecular FormulaH8O12S2Zr
InChI KeyNLOQZPHAWQDLQW-UHFFFAOYSA-J
SMILESO.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities2% max.
Catalyst support.Zirconium disulfate tetrahydrate is used in organic synthesis. It is also used in tanning white leather. Further, it is used as catalyst supports to precipitate proteins and amino acids. It is also useful in chemical analysis. It plays an essential role as an additive for lubricants and as a pigment stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only