Specifications
Chemical Name or MaterialFullerene powder
Name NoteMixed refined
CAS99685-96-8
Health Hazard 1H319-H335
Health Hazard 2GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more