Fullerene powder, hydroxylated, C{60}(OH)∣n,, Thermo Scientific Chemicals
Fullerene powder, hydroxylated, C{60}(OH)∣n,, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fullerene powder, hydroxylated, C{60}(OH)∣n,, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
Catalog number 041182.MC
also known as 041182-MC
Price (USD)/ Each
603.00
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)/ Each
603.00
Add to cart
Fullerene powder, hydroxylated, C{60}(OH)∣n,, Thermo Scientific Chemicals
Catalog number041182.MC
Price (USD)/ Each
603.00
-
Add to cart
Specifications
Chemical Name or MaterialFullerene powder
Name NoteHydroxylated, C60(OH)n
CAS99685-96-8
Health Hazard 1H228-H315-H319-H335
Health Hazard 2GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only