Thermo Scientific Chemicals
Gold Palladium wire, 0.2mm (0.008in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number: 041206.BU
25 cm, Each
Thermo Scientific Chemicals
Gold Palladium wire, 0.2mm (0.008in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number: 041206.BU
25 cm, Each
Quantity
Catalog number: 041206.BU
also known as 041206-BU
Price (USD)
Quantity
-
Have Questions?
Description
Gold Palladium wire is used as a microelectrode device, which exhibits excellent responses to hydrogen in concentrations as low as 0.5% by volume. It is also involved in the enhanced sensitivity for the detection of hydrogen.