Di-sec-butoxyaluminoxytriethoxysilane, Thermo Scientific Chemicals
Di-sec-butoxyaluminoxytriethoxysilane, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-sec-butoxyaluminoxytriethoxysilane, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 041210.09
also known as 041210-09
Price (USD)/ Each
44.80
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
44.80
Add to cart
Di-sec-butoxyaluminoxytriethoxysilane, Thermo Scientific Chemicals
Catalog number041210.09
Price (USD)/ Each
44.80
-
Add to cart
Chemical Identifiers
CAS68959-06-8
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (unspecified)Aluminum (Al): 7.5-8.5%
Assay (unspecified)Silicon (Si): 6.6-7.6%
Di-sec-butoxyaluminoxytriethoxysilane is used as Sol-gel intermediate for aluminum silicates. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only