Thermo Scientific Chemicals

Di-sec-butoxyaluminoxytriethoxysilane, Thermo Scientific Chemicals

Catalog number: 041210.30
250 g, Each
Thermo Scientific Chemicals

Di-sec-butoxyaluminoxytriethoxysilane, Thermo Scientific Chemicals

Catalog number: 041210.30
250 g, Each
Quantity
Catalog number: 041210.30
also known as 041210-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68959-06-8
Form
Liquid
Assay (unspecified)
Aluminum (Al): 7.5-8.5%
Assay (unspecified)
Silicon (Si): 6.6-7.6%

Description

Di-sec-butoxyaluminoxytriethoxysilane is used as Sol-gel intermediate for aluminum silicates. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text